Iris Scissors, OR Grade, Sklar
Iris Scissors are designed for ophthalmic procedures, these fine and sharp scissors are suitable for detailed dissection of fine tissue and fine suture removal.
- Multipurpose instrument used in various surgical environments
In addition to ophthalmic procedures, Iris scissors are used in emergency rooms and for obstetrics and gynecology as well as dermatologic procedures.
The Ready OR Grade Iris Scissors are industry standard and constructed of stainless steel.
: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
- Désignation:Iris Scissor, Premium OR-Grade
- Longueur:11.4 cm (4.5")
- Matière:Stainless steel
- Curvature:Straight
- Sterility:Non sterile
- Blade Type:Sharp/Sharp, Smooth
- Handle:Finger Rings