Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Glassware Carts, Labconco® | Avantor

Glassware Carts, Labconco®

Cart with 4 Baskets, 16×18×4", 43 lbs.
Catalog # 82006-870
Supplier:   Labconco
Fournisseur #:  8045000
Glassware Carts, Labconco®
Catalog # 82006-870
Supplier:  Labconco
Glassware Carts, Labconco®
Glassware Carts, Labconco®

Cart with 4 Baskets, 16×18×4", 43 lbs.

Price
$1,520.00
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents

Designed for use with glassware and small equipment

Carts have a welded tubular steel frame with tough, corrosion-resistant glacier white epoxy coating, four 10.2cm (4") diameter casters (two with toe locks), two handles with ergonomic extruded rubber grips, and a heavy-duty removable plastic drip pan. Interchangeable wire baskets are vinyl-coated for glassware safety and easy storage.

Ordering information: Manufacturer's lifetime warranty.

Spécifications
  • Désignation:
    Cart with 4 Baskets
  • Basket Dimensions:
    16×18×4"
  • Weight:
    43 lbs.
Accessories

Informations sur le produit

  • Catalog #
    Length
    Height
    Availability
    Price
  • Large Replacement Basket, Vinyl Coated Wire, for Cart 82006-868
    Large Replacement Basket, Vinyl Coated Wire, for Cart 82006-868
    19761-050
    81.3 cm (32")
    10.2 cm (4")
    Log in pour la disponibilité
    Each - $370.00

Informations sur le produit

  • Catalog #
    Basket Dimensions
    Length
    Availability
    Price
  • Small Replacement Basket, Vinyl Coated Wire, for Cart 82006-870
    Small Replacement Basket, Vinyl Coated Wire, for Cart 82006-870
    19761-052
    16×18×4"
    40.6 cm (16")
    Log in pour la disponibilité
    Each - $247.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.