Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Glassware Carts, Labconco® | Avantor

Glassware Carts, Labconco®

Cart with 2 Baskets, 32×18×4", 43 lbs.
Catalog # 82006-868
Supplier:   Labconco
Fournisseur #:  8032500
Glassware Carts, Labconco®
Catalog # 82006-868
Supplier:  Labconco
Glassware Carts, Labconco®
Glassware Carts, Labconco®

Cart with 2 Baskets, 32×18×4", 43 lbs.

Price
$1,520.00
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents

Designed for use with glassware and small equipment

Carts have a welded tubular steel frame with tough, corrosion-resistant glacier white epoxy coating, four 10.2cm (4") diameter casters (two with toe locks), two handles with ergonomic extruded rubber grips, and a heavy-duty removable plastic drip pan. Interchangeable wire baskets are vinyl-coated for glassware safety and easy storage.

Ordering information: Manufacturer's lifetime warranty.

Spécifications
  • Désignation:
    Cart with 2 Baskets
  • Basket Dimensions:
    32×18×4"
  • Weight:
    43 lbs.
Accessories

Informations sur le produit

  • Catalog #
    Length
    Height
    Availability
    Price
  • Large Replacement Basket, Vinyl Coated Wire, for Cart 82006-868
    Large Replacement Basket, Vinyl Coated Wire, for Cart 82006-868
    19761-050
    81.3 cm (32")
    10.2 cm (4")
    Log in pour la disponibilité
    Each - $370.00

Informations sur le produit

  • Catalog #
    Basket Dimensions
    Length
    Availability
    Price
  • Small Replacement Basket, Vinyl Coated Wire, for Cart 82006-870
    Small Replacement Basket, Vinyl Coated Wire, for Cart 82006-870
    19761-052
    16×18×4"
    40.6 cm (16")
    Log in pour la disponibilité
    Each - $247.00

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.