Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
E-C Sample Vials, WHEATON®, DWK Life Sciences

E-C Sample Vials, WHEATON®, DWK Life Sciences

20.00 ml, 28×57 mm, Non
Catalog # 80076-560
Supplier #:  W224809
E-C Sample Vials, WHEATON®, DWK Life Sciences
Catalog # 80076-560
E-C Sample Vials, WHEATON®, DWK Life Sciences
E-C Sample Vials, WHEATON®, DWK Life Sciences

20.00 ml, 28×57 mm, Non

Price
381,29 $
New Customer Promotional Price
Log in for availability
Case of 200
Product Details & Documents

Organic series clear glass vials are made from Wheaton 33® low extractable borosilicate glass.

Vials provide an inert and chemically-resistant environment for harsh, sensitive, and reactive samples. Vials accommodate size 24-400 screw caps (not included).

Certifications: Vials meet ASTM Type I Class A and USP Type I standards.

Ordering information: Packed in two shrink-wrapped packs of 100 vials.

Specifications
  • Avec bouchon:
    Non
  • Capacité:
    20.00 ml
  • Diameter ×H:
    28×57 mm