Optimize Back Pressure Regulators
Optimize Back Pressure Regulators
30 psi back pressure regulator
Catalog # 77100-074
Supplier:   Optimize
Supplier #:  10-06-00128
Optimize Back Pressure Regulators
Catalog # 77100-074
Supplier:  Optimize
Optimize Back Pressure Regulators
Optimize Back Pressure Regulators
30 psi back pressure regulator
Price
$885.89
Log in pour la disponibilité
Each
Product Details & Documents
Optimize flow-through back pressure regulators utilize an active mechanism to maintain steady back pressure across a range of mobile phase viscosities and flow rates.
  • Flow-through design
  • Utilizes an active mechanism to maintain steady back pressure across a range of conditions
  • Prevent solvent decompression and bubble formation in the flow cell
  • Reduces baseline noise and drift
  • 100 µl internal volume
Placing one downstream from your detector will prevent solvent decompression and bubble formation in the flow cell, reducing baseline noise and drift. Each pre-set Optimize stainless steel Back Pressure Regulator offers a 100 µl internal volume and includes fittings for immediate connection out of the box.
Delivery information: Includes OPTI-LOK® LP Fittings.
Spécifications
  • Désignation:
    30 psi back pressure regulator