Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
38 L (10 gal.), 46 x 30 x 30 cm (18 x 12 x 12")
Catalog # 73470-028
Supplier: Thermo Fisher Scientific
Fournisseur #: 14200-0015
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
Catalog # 73470-028
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Tanks have stepped rim for strength and overlapping skirted cover
Polypropylene tank is molded in one piece and is autoclavable
Certifications: Plastic meets USP VI and FDA 21 CFR 177.1520 standards.
Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Volume:38 L (10 gal.)
- L×W×D:46 x 30 x 30 cm (18 x 12 x 12")