Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sample Vials with Hinged Lid, Dynalon | Avantor

Sample Vials with Hinged Lid, Dynalon

35.00 ml, 31×74.5 mm, Oui
Catalog # 71070-206
Supplier:   Dynalon
Fournisseur #:  226245-50
Sample Vials with Hinged Lid, Dynalon
Catalog # 71070-206
Supplier:  Dynalon
Sample Vials with Hinged Lid, Dynalon
Sample Vials with Hinged Lid, Dynalon

35.00 ml, 31×74.5 mm, Oui

Price
$594.86
Log in pour la disponibilité
Case of 100
Product Details & Documents

General-purpose containers feature push-in friction fit hinged lids.

Lightweight vials have a wall thickness that is approximately 1 mm. LDPE construction withstands temperatures up to 80 °C (176 °F).

Spécifications
  • Avec bouchon:
    Oui
  • Capacité:
    35.00 ml
  • Diameter ×H:
    31×74.5 mm