Tissue Homogenizer, Potter-Elehjem, Ace Glass Incorporated
Potter-Elehjem Tissue Homogenizer, PTFE Pestle, 5 ml, 13 x 66 mm, 219 mm
Catalog # 71000-532
Supplier: Ace Glass
Fournisseur #: 8355-05
Jump to:
Product Details & Documents
When using this Potter-Elehjem tissue homogenizer, homogenization occurs as the sample and buffer are forced through the cylindrical portion of the mortar as the pestle is rotated downward
Cited in Biochemistry Laboratory, 2nd Edition, by Rodney Boyer, "Tools for the Preparation of Crude Cell Extracts".
The mortar is not ground, and the pestle contact surface is PTFE, threaded onto a stainless steel shaft. Tube fabricated from borosilicate glass.
Spécifications
- Capacité:5 ml
- Désignation:Potter-Elehjem Tissue Homogenizer, PTFE Pestle
- O.D.×H:13 x 66 mm
- Overall Length:219 mm
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



