Copper Wire, Arcor Electronics
Bare copper wire, 0.81 mm (0.032"), 20 AWG
66248-062
: B20
99.9% pure, soft drawn bare copper wire
In 1/4 lb. spools.
- Ø:0.81 mm (0.032")
- Désignation:Bare copper wire
- Jauge:20 AWG
99.9% pure, soft drawn bare copper wire
In 1/4 lb. spools.