pH Buffer Solution, Standardized, Lamotte | Avantor
pH Buffer Solution, Standardized, Lamotte
Standardized pH Buffer Solution, 500 ml, 7.00
Catalog # 66170-840
Fournisseur #:  2881-J
pH Buffer Solution, Standardized, Lamotte
Catalog # 66170-840
Supplier:  
CAS Number:  
pH Buffer Solution, Standardized, Lamotte
Standardized pH Buffer Solution, 500 ml, 7.00
CAS Number:  
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
BDH BUFFER REF STD PH7 COLORLESS 500ML Catalog #BDH5052-500ML
Le produit BDH5052-500ML est similaire à 66170-840 mais le fabricant et les spécifications peuvent varier.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Standardized pH buffer solutions are used for the calibration of pH meters.
Spécifications
Spécifications
  • Désignation:
    Standardized pH Buffer Solution
  • pH:
    7.00
  • Volume:
    500 ml