pH Buffer Solution, Standardized, Lamotte | Avantor
pH Buffer Solution, Standardized, Lamotte
Standardized pH Buffer Solution, 120 ml, 7.00
Catalog # 66170-838
Supplier:   LaMotte
Fournisseur #:  2881-J
pH Buffer Solution, Standardized, Lamotte
Catalog # 66170-838
Supplier:  LaMotte
CAS Number:  
pH Buffer Solution, Standardized, Lamotte
Standardized pH Buffer Solution, 120 ml, 7.00
CAS Number:  
Price
$40.82
Log in pour la disponibilité
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Standardized pH buffer solutions are used for the calibration of pH meters.
Spécifications
Spécifications
  • Désignation:
    Standardized pH Buffer Solution
  • pH:
    7.00
  • Volume:
    120 ml