Caps for 13-425 Screw-Thread Vials
Caps for 13-425 Screw-Thread Vials
Open-top screw cap, Red PTFE/white silicone, 13-425, Black, Phenolic cap
Catalog # 66065-242
Supplier:   Thermo Scientific
Supplier #:  C4015-66A
Caps for 13-425 Screw-Thread Vials
Catalog # 66065-242
Supplier:  Thermo Scientific
Caps for 13-425 Screw-Thread Vials
Open-top screw cap, Red PTFE/white silicone, 13-425, Black, Phenolic cap
Price
$231.56
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 100
Product Details & Documents
Screw thread caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs.
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Phenolic resin caps perform well at high temperatures and are compatible with exposure to corrosives
  • Caps are bonded septa resist dislodging during injection when using large diameter blunt needles
Convenient pre-assembled caps and septa minimize contamination from handling.
Spécifications
  • Taille du bouchon:
    13-425
  • Cap Type:
    Phenolic cap
  • Septum:
    Red PTFE/white silicone
  • Désignation:
    Open-top screw cap
  • Cap Color:
    Black