Caps for 13-425 Screw-Thread Vials
Open-top screw cap, Red PTFE/white silicone, 13-425, Black, Phenolic cap
Supplier: Thermo Scientific
Supplier #: C4015-66A
Jump to:
Product Details & Documents
Screw thread caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs.
- Polypropylene caps are chemically inert and suitable for most chromatography applications
- Phenolic resin caps perform well at high temperatures and are compatible with exposure to corrosives
- Caps are bonded septa resist dislodging during injection when using large diameter blunt needles
Convenient pre-assembled caps and septa minimize contamination from handling.
Spécifications
- Taille du bouchon:13-425
- Cap Type:Phenolic cap
- Septum:Red PTFE/white silicone
- Désignation:Open-top screw cap
- Cap Color:Black