Snap Caps, Wheaton
Natural cap with cross slit, 11 mm, PTFE/Silicone
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: 242775
Jump to:
Product Details & Documents
Eliminate the need for crimping and decapping tools.
- Manufactured from polyethylene
Fits the WHEATON E-Z vials® with snap ring top.
Spécifications
- Septum:PTFE/Silicone
- Désignation:Natural cap with cross slit
- Taille:11 mm