Snap Caps, Wheaton
Snap Caps, Wheaton
Natural cap with cross slit, 11 mm, PTFE/Silicone
Catalog # 66014-612
Supplier #:  242775
Snap Caps, Wheaton
Catalog # 66014-612
Snap Caps, Wheaton
Snap Caps, Wheaton
Natural cap with cross slit, 11 mm, PTFE/Silicone
Price
$1,786.00
New Customer Promotional Price
Log in for availability
Case of 1,000
Product Details & Documents
Eliminate the need for crimping and decapping tools.
  • Manufactured from polyethylene
Fits the WHEATON E-Z vials® with snap ring top.
Specifications
  • Septum:
    PTFE/Silicone
  • Désignation:
    Natural cap with cross slit
  • Taille:
    11 mm