Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
E-C Sample Vials, Glass, WHEATON®, DWK Life Sciences | Avantor

E-C Sample Vials, Glass, WHEATON®, DWK Life Sciences

4.00 ml, 15×48 mm, Clear, Oui, 13-425, Phenolic, Black, PTFE/14B rubber
Catalog # 66011-548
Fournisseur #:  224742
E-C Sample Vials, Glass, WHEATON®, DWK Life Sciences
Catalog # 66011-548
E-C Sample Vials, Glass, WHEATON®, DWK Life Sciences
E-C Sample Vials, Glass, WHEATON®, DWK Life Sciences

4.00 ml, 15×48 mm, Clear, Oui, 13-425, Phenolic, Black, PTFE/14B rubber

Price
$374.85
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 200
Product Details & Documents

Wheaton E-C Sample Vials are economically priced by offering vials without screw caps or with screw caps packaged separately.

  • Autoclavable vials and caps

Vials are offered with a choice of 14B rubber or PTFE faced 14B Rubber lined black phenolic caps. PTFE provides a totally inert inner seal and surface facing the sample or product.

Vials are offered in both clear and amber glass and are packaged in in shrink-wrapped packs of 100 vials to reduce contamination from corrugate.

Certifications: Clear vials conform to USP Type I and ASTM E438 Type 1, Class A requirements while amber vials conform to USP Type I requirements for light transmission to protect light sensitive samples.

Ordering information: Caps for vials without screw caps can be purchased separately.

Spécifications
  • Avec bouchon:
    Oui
  • Cap Material:
    Phenolic
  • Taille du bouchon:
    13-425
  • Septum:
    PTFE/14B rubber
  • Capacité:
    4.00 ml
  • Color:
    Clear
  • Diameter ×H:
    15×48 mm
  • Cap Color:
    Black
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.