Vacuum Pickup Kit | Avantor
Vacuum Pickup Kit
Vacuum pen kit, 3 star quality, includes CB-14, CB-38, CB-12
Catalog # 63043-526
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Supplier:   Excelta
Fournisseur #:  PV-10
Vacuum Pickup Kit
Catalog # 63043-526
Supplier:  Excelta
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Vacuum Pickup Kit
Vacuum Pickup Kit
Vacuum pen kit, 3 star quality, includes CB-14, CB-38, CB-12
Price
$285.85
Log in pour la disponibilité
Each
Product Details & Documents
Spécifications
  • Désignation:
    Vacuum pen kit, 3 star quality, includes CB-14, CB-38, CB-12
  • Cat. No.:
    63043-526
  • Length:
    150 mm (6")
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum cup, black Buna-N, 3 star quality, 1/8" (3.25 mm) dia. cup size
    Vacuum cup, black Buna-N, 3 star quality, 1/8" (3.25 mm) dia. cup size
    63043-266
    Log in pour la disponibilité
    Each - $7.35
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum cup, black Buna-N, 3 star quality, 3/32" (2.38 mm) dia. cup size
    Vacuum cup, black Buna-N, 3 star quality, 3/32" (2.38 mm) dia. cup size
    63043-268
    Log in pour la disponibilité
    Each - $7.55
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum cup, black Buna-N, 3 star quality, 3/4" (18.75 mm) dia. cup size
    Vacuum cup, black Buna-N, 3 star quality, 3/4" (18.75 mm) dia. cup size
    63043-270
    Log in pour la disponibilité
    Each - $15.89
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum cup, black Buna-N, 3 star quality, 3/8" (9.5 mm) dia. cup size
    Vacuum cup, black Buna-N, 3 star quality, 3/8" (9.5 mm) dia. cup size
    63043-272
    Log in pour la disponibilité
    Each - $7.47
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum cup, black Buna-N, 3 star quality, 5/8" (15.75 mm) dia. cup size
    Vacuum cup, black Buna-N, 3 star quality, 5/8" (15.75 mm) dia. cup size
    63043-274
    Log in pour la disponibilité
    Each - $15.34
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum tip, acetal, 3 star quality, 30° bent; OD 0.02", ID 0.005" cup size
    Vacuum tip, acetal, 3 star quality, 30° bent; OD 0.02", ID 0.005" cup size
    63043-610
    Log in pour la disponibilité
    Each - $212.29
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum tip, acetal, 3 star quality, 30° bent; OD 0.03", ID 0.02" cup size
    Vacuum tip, acetal, 3 star quality, 30° bent; OD 0.03", ID 0.02" cup size
    63043-612
    Log in pour la disponibilité
    Each - $186.80
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum tip, acetal, 3 star quality, 30° bent; OD 0.05", ID 0.04" cup size
    Vacuum tip, acetal, 3 star quality, 30° bent; OD 0.05", ID 0.04" cup size
    63043-614
    Log in pour la disponibilité
    Each - $179.85
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/2" (12.5 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/2" (12.5 mm) dia. cup size
    63043-616
    Log in pour la disponibilité
    Each - $13.99
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/4" (6.25 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/4" (6.25 mm) dia. cup size
    63043-618
    Log in pour la disponibilité
    Each - $13.99
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/32" (2.38 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/32" (2.38 mm) dia. cup size
    63043-622
    Log in pour la disponibilité
    Each - $14.84
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/4" (18.75 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/4" (18.75 mm) dia. cup size
    63043-624
    Log in pour la disponibilité
    Each - $25.77
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/8" (9.5 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/8" (9.5 mm) dia. cup size
    63043-626
    Log in pour la disponibilité
    Each - $13.86
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 5/8" (15.75 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 5/8" (15.75 mm) dia. cup size
    63043-628
    Log in pour la disponibilité
    Each - $25.99
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.02", ID 0.005" Cup Size
    Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.02", ID 0.005" Cup Size
    63043-644
    Log in pour la disponibilité
    Each - $207.87
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.03", ID 0.02" Cup Size
    Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.03", ID 0.02" Cup Size
    63043-646
    Log in pour la disponibilité
    Each - $179.59
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.05", ID 0.04" Cup Size
    Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.05", ID 0.04" Cup Size
    63043-648
    Log in pour la disponibilité
    Each - $199.53
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 1/2" (12.5 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 1/2" (12.5 mm) dia. cup size
    63043-650
    Log in pour la disponibilité
    Each - $13.45
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 1/8" (3.25 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 1/8" (3.25 mm) dia. cup size
    63043-654
    Log in pour la disponibilité
    Each - $14.23
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/32" (2.38 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/32" (2.38 mm) dia. cup size
    63043-656
    Log in pour la disponibilité
    Each - $13.71
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/4" (18.75 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/4" (18.75 mm) dia. cup size
    63043-658
    Log in pour la disponibilité
    Each - $25.01
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/8" (9.5 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/8" (9.5 mm) dia. cup size
    63043-660
    Log in pour la disponibilité
    Each - $13.32
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 5/8" (15.75 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 5/8" (15.75 mm) dia. cup size
    63043-662
    Log in pour la disponibilité
    Each - $23.60
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum cup, black Buna-N, 3 star quality, 3/16" (4.76 mm) dia. cup size
    Vacuum cup, black Buna-N, 3 star quality, 3/16" (4.76 mm) dia. cup size
    94024-400
    Log in pour la disponibilité
    Each - $6.23
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/16" (4.76 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/16" (4.76 mm) dia. cup size
    94024-430
    Log in pour la disponibilité
    Each - $12.26
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum replacement bulb, 2 star quality
    Vacuum replacement bulb, 2 star quality
    94024-434
    Log in pour la disponibilité
    Each - $38.18
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum cup, black Buna-N, 3 star quality, 1/2" (12.5 mm) dia. cup size
    Vacuum cup, black Buna-N, 3 star quality, 1/2" (12.5 mm) dia. cup size
    63043-262
    Log in pour la disponibilité
    Each - $7.35
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum cup, black Buna-N, 3 star quality, 1/4" (6.25 mm) dia. cup size
    Vacuum cup, black Buna-N, 3 star quality, 1/4" (6.25 mm) dia. cup size
    63043-264
    Log in pour la disponibilité
    Each - $7.10
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/8" (3.25 mm) dia. cup size
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/8" (3.25 mm) dia. cup size
    63043-620
    Log in pour la disponibilité
    Each - $13.99
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum pen only, 3 star quality
    Vacuum pen only, 3 star quality
    75880-500
    Log in pour la disponibilité
    Each - $160.78
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum systems, hand piece, 4 star quality
    Vacuum systems, hand piece, 4 star quality
    75880-688
    Log in pour la disponibilité
    Each - $267.09
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum systems, air filter, 4 star quality
    Vacuum systems, air filter, 4 star quality
    75880-690
    Log in pour la disponibilité
    Each - $33.05
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum systems, standard performance, 4 star quality
    Vacuum systems, standard performance, 4 star quality
    75880-692
    Log in pour la disponibilité
    Each - $1,213.61
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum pen only, 3 star quality
    Vacuum pen only, 3 star quality
    94024-438
    Log in pour la disponibilité
    Each - $206.06