Five-Star™ Cutters, Medium Head, Excelta | Avantor
Five-Star™ Cutters, Medium Head, Excelta
Carbon steel, 0.003 - 0.032" soft, 5¾", Medium Round Head , Long foam cushion grips, Optimum flush
Catalog # 63042-786
Five-Star™ Cutters, Medium Head, Excelta
Catalog # 63042-786
Supplier:  
Five-Star™ Cutters, Medium Head, Excelta
Five-Star™ Cutters, Medium Head, Excelta
Carbon steel, 0.003 - 0.032" soft, 5¾", Medium Round Head , Long foam cushion grips, Optimum flush
Discontinued
Product Details & Documents
Five-Star™ 7000 series medium head cutters are precision side cutters designed to make an extremely precise cut on small leads and soft wire.
  • Available either with long foam cushion grips or ESD safe molded grips
  • The dual stainless-steel leaf springs provide smooth action
These are feature with raised bearing surface, Rockwell C 63-65 hardness, and a Delrin washer for smooth operation. The CNC machining allows the joint surfaces to be perfect to allow for long life.
Spécifications
  • Matière:
    Carbon steel
  • Cutting Capacity:
    0.003 - 0.032" soft
  • Tip Style:
    Medium Round Head
  • Overall Length:
    5¾"
  • Grip Type:
    Long foam cushion grips
  • Cut Type:
    Optimum flush