Five-Star™ Cutters, Medium Head, Excelta | Avantor
Five-Star™ Cutters, Medium Head, Excelta
Carbon steel, 0.008 - 0.042" soft, 5¾", Medium Round Head , Long foam cushion grips, Semi-flush
Catalog # 63042-782
Supplier:   Excelta
Fournisseur #:  7140E
Five-Star™ Cutters, Medium Head, Excelta
Catalog # 63042-782
Supplier:  Excelta
Five-Star™ Cutters, Medium Head, Excelta
Five-Star™ Cutters, Medium Head, Excelta
Carbon steel, 0.008 - 0.042" soft, 5¾", Medium Round Head , Long foam cushion grips, Semi-flush
Price
$239.34
Log in pour la disponibilité
Each
Product Details & Documents
Five-Star™ 7000 series medium head cutters are precision side cutters designed to make an extremely precise cut on small leads and soft wire.
  • Available either with long foam cushion grips or ESD safe molded grips
  • The dual stainless-steel leaf springs provide smooth action
These are feature with raised bearing surface, Rockwell C 63-65 hardness, and a Delrin washer for smooth operation. The CNC machining allows the joint surfaces to be perfect to allow for long life.
Spécifications
  • Matière:
    Carbon steel
  • Cutting Capacity:
    0.008 - 0.042" soft
  • Tip Style:
    Medium Round Head
  • Overall Length:
    5¾"
  • Grip Type:
    Long foam cushion grips
  • Cut Type:
    Semi-flush