Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Pliers, Replaceable Tip, Straight, Excelta | Avantor

Pliers, Replaceable Tip, Straight, Excelta

Pliers, replaceable tip, 5 star quality, PTFE, 150 mm (6"), —, Straight
Catalog # 63042-564
Supplier:   Excelta
Fournisseur #:  533-US
Pliers, Replaceable Tip, Straight, Excelta
Catalog # 63042-564
Supplier:  Excelta
Pliers, Replaceable Tip, Straight, Excelta
Pliers, Replaceable Tip, Straight, Excelta

Pliers, replaceable tip, 5 star quality, PTFE, 150 mm (6"), —, Straight

Price
$550.88
Log in pour la disponibilité
Each
Product Details & Documents

These pliers are useful in straightening leads and wires when metal-to-metal contact is not desired.

  • Anti-shock trimming of leads to specific length
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use

This series of pliers offer soft jaw inserts to protect delicate parts. The jaw inserts are factory replaceable, and offered in Acetal, Brass and Nylon. The overall length is 5.75". They feature an ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs.

Spécifications
  • Désignation:
    Pliers, replaceable tip, 5 star quality
  • Longueur:
    150 mm (6")
  • Matière:
    PTFE
  • Curvature:
    Straight
  • Tip Size:
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.