Wire Stripper, Excelta
Product Details & Documents
These two tools are designed for stripping PTFE coated small wires.
- Will strip PTFE coating in one shot
- Ergonomic grips for comfort
- Double leaf springs for ease of use
These tools are made to strip small PTFE coated wire up to 30AWG. The 502-US will strip one size wire; (specify the wire gauge size by indicating the gauge at the end of the reference number. Available in 22, 24, 26, 28, and 30 AWG.) The 510E-US is a 'V' notched wire stripper which cuts the insulation on four sizes makeing stripping much easier. It features a ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. Both are made of carbon tool steel with flame hardened edges.
Specifications
- Désignation:Pliers, wire stripper, v-notch, 5 star quality, adjustable
- Longueur:137.5 mm (5.5")
- Matière:Carbon