Wire Stripper, Excelta
Wire Stripper, Excelta
Pliers, wire stripper, v-notch, 5 star quality, adjustable, Carbon, 137.5 mm (5.5")
Catalog # 63042-500
Supplier:   Excelta
Supplier #:  510E-US
Wire Stripper, Excelta
Catalog # 63042-500
Supplier:  Excelta
Wire Stripper, Excelta
Wire Stripper, Excelta
Pliers, wire stripper, v-notch, 5 star quality, adjustable, Carbon, 137.5 mm (5.5")
Price
$810.75
Log in for availability
Each
Product Details & Documents
These two tools are designed for stripping PTFE coated small wires.
  • Will strip PTFE coating in one shot
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
These tools are made to strip small PTFE coated wire up to 30AWG. The 502-US will strip one size wire; (specify the wire gauge size by indicating the gauge at the end of the reference number. Available in 22, 24, 26, 28, and 30 AWG.) The 510E-US is a 'V' notched wire stripper which cuts the insulation on four sizes makeing stripping much easier. It features a ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. Both are made of carbon tool steel with flame hardened edges.
Specifications
  • Désignation:
    Pliers, wire stripper, v-notch, 5 star quality, adjustable
  • Longueur:
    137.5 mm (5.5")
  • Matière:
    Carbon