Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Scissors, Fine, Excelta Corp®

Scissors, Fine, Excelta Corp®

Scissors, long blade, 2 star quality, 4.5" (112.5 mm), Stainless steel AISI 440, 44 mm (1¾"), Straight
Catalog # 63041-842
Supplier:   Excelta
Supplier #:  298A
Scissors, Fine, Excelta Corp®
Catalog # 63041-842
Supplier:  Excelta
Scissors, Fine, Excelta Corp®
Scissors, Fine, Excelta Corp®

Scissors, long blade, 2 star quality, 4.5" (112.5 mm), Stainless steel AISI 440, 44 mm (1¾"), Straight

Price
$25.11
Log in for availability
Each
Product Details & Documents

These high quality scissors are designed for delicate work and close tolerance cutting.

  • Cleanroom safe, stainless steel
  • Variety of blade lengths

These stainless steel scissors offer fine blades for clean cutting. They are all made of AISI 440 stainless steel. They are offered in a variety of blade lengths.

Specifications
  • Désignation:
    Scissors, long blade, 2 star quality
  • Longueur:
    4.5" (112.5 mm)
  • Matière:
    Stainless steel AISI 440
  • Curvature:
    Straight
  • Blade Length:
    44 mm (1¾")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.