Scissors, Fine, Excelta Corp®
Scissors, long blade, 2 star quality, 3.5" (87.5 mm), Stainless steel AISI 440, 31 mm (1¼"), Straight
  63041-838
 :   Excelta
 :  297
Scissors, Fine, Excelta Corp®
  63041-838
 :  Excelta
Scissors, Fine, Excelta Corp®
Scissors, Fine, Excelta Corp®
Scissors, long blade, 2 star quality, 3.5" (87.5 mm), Stainless steel AISI 440, 31 mm (1¼"), Straight
$17.68
Each
These high quality scissors are designed for delicate work and close tolerance cutting.
  • Cleanroom safe, stainless steel
  • Variety of blade lengths
These stainless steel scissors offer fine blades for clean cutting. They are all made of AISI 440 stainless steel. They are offered in a variety of blade lengths.
  • Désignation:
    Scissors, long blade, 2 star quality
  • Longueur:
    3.5" (87.5 mm)
  • Matière:
    Stainless steel AISI 440
  • Curvature:
    Straight
  • Blade Length:
    31 mm (1¼")