Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Scissors, Fine, Excelta Corp® | Avantor

Scissors, Fine, Excelta Corp®

Scissors, medium fine blade, 2 star quality, 3.75" (93.75 mm), Stainless steel AISI 440, 25.4 mm (1"), 20° curved
Catalog # 63041-834
Supplier:   Excelta
Fournisseur #:  291
Scissors, Fine, Excelta Corp®
Catalog # 63041-834
Supplier:  Excelta
Scissors, Fine, Excelta Corp®
Scissors, Fine, Excelta Corp®

Scissors, medium fine blade, 2 star quality, 3.75" (93.75 mm), Stainless steel AISI 440, 25.4 mm (1"), 20° curved

Price
22,67 $
Log in pour la disponibilité
Each
Product Details & Documents

These high quality scissors are designed for delicate work and close tolerance cutting.

  • Cleanroom safe, stainless steel
  • Variety of blade lengths

These stainless steel scissors offer fine blades for clean cutting. They are all made of AISI 440 stainless steel. They are offered in a variety of blade lengths.

Spécifications
  • Désignation:
    Scissors, medium fine blade, 2 star quality
  • Longueur:
    3.75" (93.75 mm)
  • Matière:
    Stainless steel AISI 440
  • Curvature:
    20° curved
  • Blade Length:
    25.4 mm (1")
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.