Scissors, Fine, Excelta Corp®
Scissors, medium fine blade, 2 star quality, 3.75" (93.75 mm), Stainless steel AISI 440, 25.4 mm (1"), Straight
Catalog # 63041-832
Supplier:   Excelta
 :  290
Scissors, Fine, Excelta Corp®
Catalog # 63041-832
Supplier:  Excelta
Scissors, Fine, Excelta Corp®
Scissors, Fine, Excelta Corp®
Scissors, medium fine blade, 2 star quality, 3.75" (93.75 mm), Stainless steel AISI 440, 25.4 mm (1"), Straight
Price
$22.44
Log in pour la disponibilité
Each
Product Details & Documents
These high quality scissors are designed for delicate work and close tolerance cutting.
  • Cleanroom safe, stainless steel
  • Variety of blade lengths
These stainless steel scissors offer fine blades for clean cutting. They are all made of AISI 440 stainless steel. They are offered in a variety of blade lengths.
Spécifications
  • Désignation:
    Scissors, medium fine blade, 2 star quality
  • Longueur:
    3.75" (93.75 mm)
  • Matière:
    Stainless steel AISI 440
  • Curvature:
    Straight
  • Blade Length:
    25.4 mm (1")