Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Scissors, Fine, Excelta Corp® | Avantor

Scissors, Fine, Excelta Corp®

Scissors, short fine blade, 2 star quality, 3.25" (81.25 mm), Stainless steel AISI 440, 18.75 mm (0.75"), Straight
Catalog # 63041-826
Supplier:   Excelta
Fournisseur #:  288
Scissors, Fine, Excelta Corp®
Catalog # 63041-826
Supplier:  Excelta
Scissors, Fine, Excelta Corp®
Scissors, Fine, Excelta Corp®

Scissors, short fine blade, 2 star quality, 3.25" (81.25 mm), Stainless steel AISI 440, 18.75 mm (0.75"), Straight

Price
$22.89
Log in pour la disponibilité
Each
Product Details & Documents

These high quality scissors are designed for delicate work and close tolerance cutting.

  • Cleanroom safe, stainless steel
  • Variety of blade lengths

These stainless steel scissors offer fine blades for clean cutting. They are all made of AISI 440 stainless steel. They are offered in a variety of blade lengths.

Spécifications
  • Désignation:
    Scissors, short fine blade, 2 star quality
  • Longueur:
    3.25" (81.25 mm)
  • Matière:
    Stainless steel AISI 440
  • Curvature:
    Straight
  • Blade Length:
    18.75 mm (0.75")
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.