Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Scissors, Fine, Excelta Corp®

Scissors, Fine, Excelta Corp®

Scissors, slim blade, 2 star quality, 4.75" (118.75 mm), Stainless steel, 31 mm (1¼"), Straight
Catalog # 63041-752
Supplier:   Excelta
Supplier #:  271
Scissors, Fine, Excelta Corp®
Catalog # 63041-752
Supplier:  Excelta
Scissors, Fine, Excelta Corp®
Scissors, Fine, Excelta Corp®

Scissors, slim blade, 2 star quality, 4.75" (118.75 mm), Stainless steel, 31 mm (1¼"), Straight

Price
$26.86
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

These high quality scissors are designed for delicate work and close tolerance cutting.

  • Cleanroom safe, stainless steel
  • Variety of blade lengths

These stainless steel scissors offer fine blades for clean cutting. They are all made of AISI 440 stainless steel. They are offered in a variety of blade lengths.

Specifications
  • Désignation:
    Scissors, slim blade, 2 star quality
  • Longueur:
    4.75" (118.75 mm)
  • Matière:
    Stainless steel
  • Curvature:
    Straight
  • Blade Length:
    31 mm (1¼")