Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Replacement tip for 268, 2 star quality, Nylon, 0.13" (3.25 mm)
Catalog # 63041-744
Supplier: Excelta
Fournisseur #: 268A
Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Catalog # 63041-744
Supplier: Excelta
Jump to:
Product Details & Documents
Spécifications
- Bristle Material:Nylon
- Désignation:Replacement tip for 268, 2 star quality
- Brush Head Width:0.13" (3.25 mm)