Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Replacement tip for 266, 2 star quality, Steel, 0.13" (3.25 mm)
Catalog # 63041-736
Supplier: Excelta
: 266A
Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Catalog # 63041-736
Supplier: Excelta
Jump to:
Product Details & Documents
Spécifications
- Bristle Material:Steel
- Désignation:Replacement tip for 266, 2 star quality
- Brush Head Width:0.13" (3.25 mm)