Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Replacement tip for 264, 2 star quality, Fiberglass, 0.05" (1.25 mm)
63041-728
: Excelta
: 264A
- Bristle Material:Fiberglass
- Désignation:Replacement tip for 264, 2 star quality
- Brush Head Width:0.05" (1.25 mm)