Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Replacement tip for 263, 2 star quality, Fiberglass, 0.32" (8 mm)
Catalog # 63041-712
Supplier: Excelta
Fournisseur #: 263A
Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Catalog # 63041-712
Supplier: Excelta
Jump to:
Product Details & Documents
Spécifications
- Bristle Material:Fiberglass
- Désignation:Replacement tip for 263, 2 star quality
- Brush Head Width:0.32" (8 mm)