PYREX® Drying Trays, Corning | Avantor
PYREX® Drying Trays, Corning
3500 ml, 34.9 x 24.9 x 6 cm (1323/32 x 913/16 x 211/32")
Catalog # 62670-016
Supplier:   Corning
Fournisseur #:  3175-10
PYREX® Drying Trays, Corning
Catalog # 62670-016
Supplier:  Corning
PYREX® Drying Trays, Corning
PYREX® Drying Trays, Corning
3500 ml, 34.9 x 24.9 x 6 cm (1323/32 x 913/16 x 211/32")
Price
$226.00
Log in pour la disponibilité
Case of 4
Product Details & Documents
These dishes made of PYREX® borosilicate glass will withstand hot air or steam sterilization and are useful for drying, staining or developing trays.
  • Excellent chemical resistance
  • Good temperature resistance
  • Easy to clean
They can be used with gel electrophoresis or paper chromatography.
Caution: For oven use only. Do not use over direct heat.
Spécifications
  • Volume:
    3500 ml
  • L×W×H:
    34.9 x 24.9 x 6 cm (1323/32 x 913/16 x 211/32")