Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Trypsinizing Flasks with Pourout and Caps, Wheaton | Avantor

Trypsinizing Flasks with Pourout and Caps, Wheaton

1 L, 700 ml, 51-400
Catalog # 62401-656
Fournisseur #:  355757
Trypsinizing Flasks with Pourout and Caps, Wheaton
Catalog # 62401-656
Trypsinizing Flasks with Pourout and Caps, Wheaton
Trypsinizing Flasks with Pourout and Caps, Wheaton

1 L, 700 ml, 51-400

Price
$291.20
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 1
Product Details & Documents

For trypsinizing and decanting cells, the flask is designed for use with magnetic stirrers and spin bars

Made from Wheaton 33® low extractable borosilicate glass flasks with deep fluted sides to promote turbulence. Special internal dam under the sidearm retains untrypsinized particles when decanting. A deep-skirted 20-415 screw cap on the sidearm prevents contamination of flask contents. Screw caps have white styrene butadiene rubber liners.

Ordering information: Supplied with caps.

Spécifications
  • Taille du bouchon:
    51-400
  • Capacité:
    1 L
  • Working Volume:
    700 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.