White Large Display Timer, Sper Scientific
White Large Display Timer, Sper Scientific
White large display timer, certified, 5×3¾×½", 6.7 oz.
Catalog # 62364-205
Supplier:   Sper Scientific
Supplier #:  810005C
White Large Display Timer, Sper Scientific
Catalog # 62364-205
Supplier:  Sper Scientific
White Large Display Timer, Sper Scientific
White Large Display Timer, Sper Scientific
White large display timer, certified, 5×3¾×½", 6.7 oz.
Price
$284.98
Log in for availability
Each
Product Details & Documents
This easy-to-read, large display timer features the largest display available on a timer: 3×2¹/₂"
  • Can be easily read from across a room
  • White case has extra large start/stop button and a loud alarm
  • Wall mount or fold for bench top use
  • Runs for approximately two years on one AAA battery
Simultaneously displays either one count up channel and a clock, or two count down channels. All channels have a range of 24 hours with 1 second resolution. Also indicates how much time has elapsed after a count down alarm sounds and conveniently recalls the last timer setting for reuse.
Certifications: N.I.S.T. traceable certificate of compliance for certified option.
Ordering information: One AAA battery included. Five Year Warranty.
Specifications
  • Désignation:
    White large display timer, certified
  • W×H×D:
    5×3¾×½"
  • Weight:
    6.7 oz.