Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
Adaptable to a wide range of applications, these recorders feature a 15.2cm (6") readable chart, AC power with battery backup, user-selectable recording times and temperature ranges, and a remote probe
Increased shielding eliminates electronic noise interference and allows smoother pen movement.
Ordering information: Recorders include red pen, AC adapter, 9V battery, and 1.2m (4') bead wire probe. Charts must be purchased separately.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
- Plage de température:–50...50 °C/°F
- Désignation:24-Hour Chart