Data-Trend Multipurpose Temperature Chart Recorders, Dickson® | Avantor
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
24-Hour Chart, 50...100 °C/°F
Catalog # 61280-561
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
Catalog # 61280-561
Supplier:  
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
24-Hour Chart, 50...100 °C/°F
Discontinued
Product Details & Documents
Adaptable to a wide range of applications, these recorders feature a 15.2cm (6") readable chart, AC power with battery backup, user-selectable recording times and temperature ranges, and a remote probe
Increased shielding eliminates electronic noise interference and allows smoother pen movement.
Ordering information: Recorders include red pen, AC adapter, 9V battery, and 1.2m (4') bead wire probe. Charts must be purchased separately.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Spécifications
  • Plage de température:
    50...100 °C/°F
  • Désignation:
    24-Hour Chart
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Red Pens
    Red Pens
    61280-132
    Log in pour la disponibilité
    Pack of 6 - $244.14