Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
24-Hour Chart, 32...100 °F
Catalog # 61280-557
Supplier:   Dickson
Supplier #:  C658
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
Catalog # 61280-557
Supplier:  Dickson
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
Data-Trend Multipurpose Temperature/Humidity Chart Recorders, Dickson®
24-Hour Chart, 32...100 °F
Price
$192.29
Log in for availability
Pack of 60
Product Details & Documents
Adaptable to a wide range of applications, these recorders feature a 15.2cm (6") readable chart, AC power with battery backup, and user-selectable recording times and temperature ranges
Increased shielding eliminates electronic noise interference and allows smoother pen movement.
Ordering information: Recorders include one red pen, one blue pen, AC adapter, and 9V battery. Charts must be purchased separately.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Specifications
  • Plage de température:
    32...100 °F
  • Désignation:
    24-Hour Chart
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • NIST Traceable Calibration 1-Point Service
    NIST Traceable Calibration 1-Point Service
    13450-754
    Log in for availability
    Each - $291.26
Product Information
  • Catalog #
    Availability
    Price
  • NIST & A2LA Traceable Calibration 3-Point Service
    NIST & A2LA Traceable Calibration 3-Point Service
    13450-756
    Log in for availability
    Each - $848.52
Product Information
  • Catalog #
    Availability
    Price
  • Pens, Three Red and Three Blue
    Pens, Three Red and Three Blue
    61280-134
    Log in for availability
    Pack of 6 - $244.14