TH8 Remote Sensing Temperature/Humidity/Dew Point Recorders, Dickson®
TH8 Remote Sensing Temperature/Humidity/Dew Point Recorders, Dickson®
24-Hour Chart, –20...50 °C
Catalog # 61280-281
Supplier:   Dickson
Supplier #:  C472
TH8 Remote Sensing Temperature/Humidity/Dew Point Recorders, Dickson®
Catalog # 61280-281
Supplier:  Dickson
TH8 Remote Sensing Temperature/Humidity/Dew Point Recorders, Dickson®
TH8 Remote Sensing Temperature/Humidity/Dew Point Recorders, Dickson®
24-Hour Chart, –20...50 °C
Price
$196.06
Log in pour la disponibilité
Pack of 60
Product Details & Documents
Units are ideal for remote sensing of temperature and humidity in refrigerators, coolers, cleanrooms, chambers, storage rooms, critical equipment storage, tight locations, and for monitoring IAQ and human comfort
Units 61279-932 and 61279-934 feature 2.5cm (1") 3.5 digit LCD, viewable from 7.6m (25'). Unit 61279-934 features an audio/visual alarm and a remote temperature/relative humidity probe with a 3m (10') cord. All units are portable or wall-mountable.
Recorders have a rugged, compact enclosure with an ABS case, polycarbonate door, and aluminum dial. The large 20.3cm (8") chart offers greater resolution. The flip-up pen arm allows for easy chart and pen changes. Units feature user-selectable temperature ranges and recording times, and 9V battery backup.
Ordering information: Units 61279-932 and -934 are supplied with 120V AC adapter and 9V battery for backup power. Unit 14905-000 is battery operated and is supplied with four AA batteries. All units include pens. Charts not included.
Spécifications
  • Plage de température:
    –20...50 °C
  • Désignation:
    24-Hour Chart
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • NIST Traceable Calibration 1-Point Service
    NIST Traceable Calibration 1-Point Service
    13450-754
    Log in pour la disponibilité
    Each - $291.26
Informations sur le produit
  • Catalog #
    Availability
    Price
  • NIST & A2LA Traceable Calibration 3-Point Service
    NIST & A2LA Traceable Calibration 3-Point Service
    13450-756
    Log in pour la disponibilité
    Each - $848.52
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Pens, Three Red and Three Blue
    Pens, Three Red and Three Blue
    61280-134
    Log in pour la disponibilité
    Pack of 6 - $244.14