Temperature Chart Recorders, Dickson®
7-Day Chart, 0...250
  61280-237
Temperature Chart Recorders, Dickson®
  61280-237
 :  
Temperature Chart Recorders, Dickson®
Temperature Chart Recorders, Dickson®
7-Day Chart, 0...250
Recorders are ideal for monitoring long-term temperature trends in areas where proper temperatures are critical to product stability.
  • Cord-free, portable, and easy to use
  • Recorders can be placed directly in an environment that needs to be monitored, and mounted on a desk or wall
  • Feature spiral-wound bimetallic strip transducer, hinged door, built-in slot for optional tie-down cable and lock, battery door
  • Durable, corrosion-resistant black ABS and polycarbonate enclosure
  • Recorders come with or without digital LCD, and can be set to 7-day or 24-hour recording times
  • °C/°F switchable
Designed for use in blood banks, laboratories, storage areas, refrigerators, or freezers.
Average response time: 11 min. to move 63% of full scale
 : A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology. CE marked. IP21 rated.
 : Each recorder comes with a AA alkaline battery and a red pen. Charts must be purchased separately.
 : VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
  • Plage de température:
    0...250
  • Désignation:
    7-Day Chart
Accessories
  •  
    Description
  • 61280-132
    Red Pens
    Pack of 6 - $244.14
  •  
    Description
  • 13450-754
    NIST Traceable Calibration 1-Point Service
    Each - $291.26
  •  
    Description
  • 13450-756
    NIST & A2LA Traceable Calibration 3-Point Service
    Each - $848.52