SC3 Super Compact Temperature Recorders, Dickson®
SC3 Super Compact Temperature Recorders, Dickson®
–15...10 °C (4...50 °F)
Catalog # 61280-040
Small Business Enterprise
Supplier:   Dickson
Supplier #:  C312
SC3 Super Compact Temperature Recorders, Dickson®
Catalog # 61280-040
Supplier:  Dickson
Small Business Enterprise
SC3 Super Compact Temperature Recorders, Dickson®
SC3 Super Compact Temperature Recorders, Dickson®
–15...10 °C (4...50 °F)
Price
$188.58
Log in pour la disponibilité
Each
Product Details & Documents
Reusable and compact recorders are built with a rugged case to withstand the rigors of transportation and storage applications
Economical recorders feature easy user calibration and switchable control of temperature, °C/°F, and time, 24 hours/7 days. Designed with specialized slots for unit security. Optional tamper seals can be used to ensure the integrity of the chart. Units can be wall-mounted, suspended, or used free-standing, and work with charts that have a diameter of 75mm (3").
Ordering information: Supplied with one AA battery, pen, and instruction manual.
Spécifications
  • Plage de température:
    –15...10 °C (4...50 °F)
  • Cat. No.:
    61280-040
  • Accuracy:
  • Dimensions:
  • Weight:
Recommendations