VWR® Traceable® Jumbo Timer
VWR® Traceable® Jumbo Timer
White, 9½×14×1.3 cm (3¾×5½×½")
Catalog # 61161-346
VWR® Traceable® Jumbo Timer
Catalog # 61161-346
Supplier:  
VWR® Traceable® Jumbo Timer
VWR® Traceable® Jumbo Timer
White, 9½×14×1.3 cm (3¾×5½×½")
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
TIMER LRG. DISPLAY WHITE Catalog #62370-068
Product 62370-068 is similar to 61161-346 but manufacturer and specifications may vary.
VWR TIMER TRACEABLE 2-MEMORY Catalog #62344-908
Product 62344-908 is similar to 61161-346 but manufacturer and specifications may vary.
Product Details & Documents
Ideal for catching test results, signaling time intervals, and alerting appointments.
  • Traceable®
  • Two-channel display
  • Countdown alarm, stopwatch, and time-of-day clock
Alarm timer features large, 2.5 cm (1") digits for 'across the room' readability. It simultaneously maintains two separate countdown timing channels with distinct alarms, stopwatch, and time-of-day options. Channels may be programmed individually or simultaneously. Count up or down from 23 hours, 59 minutes, 59 seconds. Alarm may be turned off manually or silences automatically after one minute. Just touch a button to return the display to the last programmed time for repeated testing applications.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a two-year AAA alkaline battery.
Specifications
  • Désignation:
    White
  • W×H×D:
    9½×14×1.3 cm (3¾×5½×½")