Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
8 L (2 gal.), 20 x 20 x 20 cm (8 x 8 x 8")
Catalog # 60463-166
Supplier: Thermo Fisher Scientific
Fournisseur #: 14100-0002
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Catalog # 60463-166
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Tanks have stepped rim for strength and overlapping skirted cover
Molded in one piece with good chemical resistance
Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Spécifications
- Volume:8 L (2 gal.)
- L×W×D:20 x 20 x 20 cm (8 x 8 x 8")