Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
50 L (13 gal.), 61 x 30 x 30 cm (24 x 12 x 12")
Catalog # 60463-142
Supplier: Thermo Fisher Scientific
Supplier #: 14100-0020
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Catalog # 60463-142
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Tanks have stepped rim for strength and overlapping skirted cover
Molded in one piece with good chemical resistance
Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Spécifications
- Volume:50 L (13 gal.)
- L×W×D:61 x 30 x 30 cm (24 x 12 x 12")