Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
8 L (2 gal.), 20 x 20 x 20 cm (8 x 8 x 8")
Catalog # 60463-105
Supplier: Thermo Fisher Scientific
Supplier #: 14200-0002
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
Catalog # 60463-105
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Tanks have stepped rim for strength and overlapping skirted cover
Polypropylene tank is molded in one piece and is autoclavable
Certifications: Plastic meets USP VI and FDA 21 CFR 177.1520 standards.
Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Volume:8 L (2 gal.)
- L×W×D:20 x 20 x 20 cm (8 x 8 x 8")