Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
30 L (8 gal.), 30 x 30 x 30 cm (12 x 12 x 12")
  60463-104
 :  14200-0010
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
  60463-104
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
30 L (8 gal.), 30 x 30 x 30 cm (12 x 12 x 12")
$589.60
Each
Tanks have stepped rim for strength and overlapping skirted cover
Polypropylene tank is molded in one piece and is autoclavable
 : Plastic meets USP VI and FDA 21 CFR 177.1520 standards.
 : Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
  • Volume:
    30 L (8 gal.)
  • L×W×D:
    30 x 30 x 30 cm (12 x 12 x 12")