Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
160 L (40 gal.), 61 x 45 x 60 cm (24 x 18 x 24")
Catalog # 60463-095
Supplier: Thermo Fisher Scientific
Fournisseur #: 14100-0065
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Catalog # 60463-095
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Tanks have stepped rim for strength and overlapping skirted cover
Molded in one piece with good chemical resistance
Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Spécifications
- Volume:160 L (40 gal.)
- L×W×D:61 x 45 x 60 cm (24 x 18 x 24")