Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
120 L (32 gal.), 61 x 45 x 45 cm (24 x 18 x 18")
Catalog # 60463-084
Supplier: Thermo Fisher Scientific
Fournisseur #: 14100-0045
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Catalog # 60463-084
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Tanks have stepped rim for strength and overlapping skirted cover
Molded in one piece with good chemical resistance
: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Spécifications
- Volume:120 L (32 gal.)
- L×W×D:61 x 45 x 45 cm (24 x 18 x 18")