Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific | Avantor

Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific

60 L (16 gal.), 45 x 30 x 45 cm (18 x 12 x 18")
Catalog # 60463-062
Fournisseur #:  14100-0040
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Catalog # 60463-062
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific

60 L (16 gal.), 45 x 30 x 45 cm (18 x 12 x 18")

Price
627,20 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents

Tanks have stepped rim for strength and overlapping skirted cover

Molded in one piece with good chemical resistance

Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.

Spécifications
  • Volume:
    60 L (16 gal.)
  • L×W×D:
    45 x 30 x 45 cm (18 x 12 x 18")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.