Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
20 L (5 gal.), 25 x 25 x 36 cm (10 x 10 x 14")
Catalog # 60463-030
Supplier: Thermo Fisher Scientific
: 14100-0005
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Catalog # 60463-030
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Tanks have stepped rim for strength and overlapping skirted cover
Molded in one piece with good chemical resistance
: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Spécifications
- Volume:20 L (5 gal.)
- L×W×D:25 x 25 x 36 cm (10 x 10 x 14")