VWR® Talon® Large Lab-Frames | Avantor
VWR® Talon® Large Lab-Frames
Lab-Frame with Aluminum Rods, Aluminum
Catalog # 60077-003
Supplier:   VWR International
VWR® Talon® Large Lab-Frames
Catalog # 60077-003
Supplier:  VWR International
VWR® Talon® Large Lab-Frames
VWR® Talon® Large Lab-Frames
Lab-Frame with Aluminum Rods, Aluminum
Price
$3,582.92
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
These 1220x1220mm (48x48") frames are ideal for complex glassware setups.
  • For large-capacity setups
  • Stable
  • Ideal for complex glass setups
Base is 460mm (18") wide for stability.
Ordering information: Each Lab Frame includes ten 51 mm (2") rods, three 457 mm (18") rods, two 914 mm (36") rods, ten 1220 mm (48") rods, four rod end connectors, thirty-eight S-connectors, and six Lab Frame feet.
Spécifications
  • Désignation:
    Lab-Frame with Aluminum Rods
  • Matière:
    Aluminum