Synthware Replacement T-Bore Glass Stopcocks
Replacement T-bore glass stopcock, 6 mm
  60005-184
 :  S512044
Synthware Replacement T-Bore Glass Stopcocks
  60005-184
Synthware Replacement T-Bore Glass Stopcocks
Synthware Replacement T-Bore Glass Stopcocks
Replacement T-bore glass stopcock, 6 mm
$60.18
Each
Glass plug ONLY.
  • Heavy wall construction
  • 33 expansion borosilicate glass
  • Standard ground glass joints work with other brands
For use with Kemtech S69 series stainless steel retention clips.
  • Désignation:
    Replacement T-bore glass stopcock
  • Bore Size:
    6 mm