Synthware Replacement T-Bore Glass Stopcocks
Replacement T-bore glass stopcock, 6 mm
60005-184
: S512044
Glass plug ONLY.
- Heavy wall construction
- 33 expansion borosilicate glass
- Standard ground glass joints work with other brands
For use with Kemtech S69 series stainless steel retention clips.
- Désignation:Replacement T-bore glass stopcock
- Bore Size:6 mm